Paraplast Plus®, pellets for electron microscopy (fast-melting), Sigma-Aldrich®
Catalog # 76258-1KG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Melting Pt:56...57 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:232-315-6
CAS Number:125387-89-5
:8012-95-1
MDL Number:MFCD00131795