Order Entry
Belgium
ContactUsLinkComponent
 
Tungsten Standard Solution, 1000 mg/L W in Water (from W), Standard for ICP
  ACSDC066.W.L1
 :  CPACHEM
New Product
 :  
Tungsten Standard Solution, 1000 mg/L W in Water (from W), Standard for ICP
  ACSDC066.W.L1
 :  CPACHEM
New Product
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Element:
    Tungsten
  • Symbol:
    W

 

 

Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.

 : ISO 17034, 17025, 9001 certified.