ESD Safety Shoes, Lacing System, Stefano XXSG Boa® GTX Low S3
Supplier: ELTEN
New Product
Certificates
About this item
ESD safety shoe made from hydrophobized cowhide leather upper and a PU/PU sole (Wellmaxx Safety Grip).
- All weather Gore-Tex performance membrane
- Full, padded bellows tongue
- Full-length inlay sole ARTISAN ESD
- Boa® fit lacing system
- Non-metallic, penetration-resistant midsole
- Steel toe cap
- PU toe protection
Indoors and outdoors, areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black/blue
Certifications: EN ISO 20345 S3 CI/SRC, form A.
Specifications
- Catalog No:
- ELTN728751/44
- ELTN728751/36
- ELTN728751/45
- ELTN728751/39
- ELTN728751/37
- ELTN728751/46
- ELTN728751/40
- ELTN728751/38
- ELTN728751/47
- ELTN728751/41
- ELTN728751/48
- ELTN728751/42
- ELTN728751/43
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001