ESD Safety Ankle Boots, Lace-Up, Ian XXTP Pro Mid S3 S
Supplier: ELTEN
New Product
Certificates
About this item
ESD safety ankle boots made from hydrophobised microfibre/hydrophobised textile material and a TPU/PU sole (Wellmaxx Trainers Power).
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO black
- Non-metallic, penetration-resistant midsole
- Composite toe cap
- TPU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Grey
Certifications: EN ISO 20345 S3S FO/SR/SC, form B.
Specifications
- Catalog No:
- ELTN761321/39
- ELTN761321/47
- ELTN761321/40
- ELTN761321/35
- ELTN761321/36
- ELTN761321/41
- ELTN761321/37
- ELTN761321/48
- ELTN761321/42
- ELTN761321/49
- ELTN761321/43
- ELTN761321/44
- ELTN761321/45
- ELTN761321/46
- ELTN761321/38
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001