ESD Safety Ankle Boots, Lacing Sytem, Ian XXTP Pro Boa® Mid S3 S
: ELTEN
ESD safety ankle boots made from hydrophobised microfibre/hydrophobised textile upper and a TPU/PU sole (Wellmaxx Trainers Power).
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO black
- Boa® fit lacing system
- Non-metallic, penetration-resistant midsole
- Composite toe cap
- TPU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Blue
: EN ISO 20345 S3S FO/SR/SC, form B.
- Catalog No:
- ELTN761331/45
- ELTN761331/46
- ELTN761331/47
- ELTN761331/48
- ELTN761331/37
- ELTN761331/49
- ELTN761331/38
- ELTN761331/39
- ELTN761331/35
- ELTN761331/40
- ELTN761331/36
- ELTN761331/41
- ELTN761331/43
- ELTN761331/44
- ELTN761331/42
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together