ContactUsLinkComponent
BizTex® Microporous Coveralls, Type 3/4/5/6
BizTex® Microporous Coveralls, Type 3/4/5/6
Supplier:  Portwest
Certificates
undefined
BizTex® Microporous Coveralls, Type 3/4/5/6
Supplier:  Portwest
Specifications
  • Catalog No:
  • PRTWST70YERXL
  • PRTWST70YERXXL
  • PRTWST70YERM
  • PRTWST70YERL
  • Garment application:
  • Chemical protection
  • Chemical protection
  • Chemical protection
  • Chemical protection
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • BizTex® micro plus
  • BizTex® micro plus
  • BizTex® micro plus
  • BizTex® micro plus
  • Weight:
  • 82 g
  • 82 g
  • 82 g
  • 82 g