Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ UV375)
Supplier: AAT BIOQUEST
Certificates
About this item
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: mFluor™ UV375
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB102800X1
- AATB102800X0
- Antigen symbol:
- CD28
- CD28
- Antigen name:
- CD28 antigen
- CD28 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- mFluor™ UV375
- mFluor™ UV375
- Clone:
- CD28.2
- CD28.2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure, do not freeze
- 2...8 °C with minimised light exposure, do not freeze
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together