ContactUsLinkComponent
Assistent Brush with Nylon Bristles
Assistent Brush with Nylon Bristles
Catalog # HECH91120000258
Assistent Brush with Nylon Bristles
Catalog # HECH91120000258
Specifications
  • Bristle material:
    Nylon
  • Brush head Ø:
    8 mm
  • Handle material:
    Stainless steel
  • Total length:
    250 mm
  • Pk:
    10
Specifications
About this item
Brush with nylon bristles for use with laboratory glassware.
  • Black bonded cap
  • Stainless steel handle