Anti-VOPP1 Rabbit Polyclonal Antibody
Catalog # PRSI55-698
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Vesicular, overexpressed in cancer, prosurvival protein 1
- Antigen symbol:VOPP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino Acid Range = 65-95
- Form:Liquid
- Gene ID:Q96AW1
- Antigen synonyms:VOPP1
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:19 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ECOP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 65-95 amino acids from the Central region of human ECOP.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
Increases the transcriptional activity of NFKB1 by facilitating its nuclear translocation, DNA-binding and associated apoptotic response, when overexpressed.
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: VOPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse