Anti-ZNF674 Rabbit Polyclonal Antibody
PRSI55-504
New Product
- Antibody type:Primary
- Antigen name:Zinc finger protein 674
- Antigen symbol:ZNF674
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino Acid Range = 74-102
- Form:Liquid
- Gene ID:Q2M3X9
- Antigen synonyms:ZNF674
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:67 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ZNF674 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 74-102 amino acids from the N-terminal region of human ZNF674.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
This gene encodes a zinc finger protein with an N-terminal Kruppel-associated box-containing (KRAB) domain and 11 Kruppel-type C2H2 zinc finger domains. Like other zinc finger proteins, this gene may function as a transcription factor. This gene resides on an area of chromosome X that has been implicated in nonsyndromic X-linked mental retardation. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq].
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ZNF674
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human