Anti-CLEC2L Rabbit Polyclonal Antibody
PRSI55-503
New Product
- Antibody type:Primary
- Antigen name:C-type lectin domain family 2 member L
- Antigen symbol:CLEC2L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Amino Acid Range = 26-54
- Form:Liquid
- Gene ID:P0C7M8
- Antigen synonyms:CLEC2L
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:24 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CLEC2L antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human CLEC2L.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
For WB starting dilution is: 1:1000
Type: Primary
Antigen: CLEC2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse