Order Entry
Belgium
ContactUsLinkComponent
 
Anti-MYC Recombinant Antibody [Clone: 9E10]
  PRSI24-054
New Product
Anti-MYC Recombinant Antibody [Clone: 9E10]
  PRSI24-054
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    MYC proto-oncogene, bHLH transcription factor
  • Antigen symbol:
    MYC
  • Clonality:
    Recombinant
  • Clone:
    9E10
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2c kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    4609
  • Antigen synonyms:
    cmyc|Class E basic helix-loop-helix protein 39|Proto-oncogene c-Myc|BHLHE39|Transcription factor p64
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    A synthetic peptide corresponding to aa 408-439 from C-terminus of human c-myc.
  • Size:
    0,2 mg
  • Pack type:
    Vial
  • Pk:
    0,2 mg

 

 

Type: Primary
Antigen: MYC proto-oncogene, bHLH transcription factor
Clonality: Recombinant
Clone: 9E10
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2c kappa
Reactivity: Human