Spécifications
- Antibody type:Primary
- Antigen name:arginase 1
- Antigen symbol:ARG1
- Clonality:Monoclonal
- Clone:ARG1/1126
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P05089
- Antigen synonyms:ARG1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:A recombinant fragment from amino acids 1-150 of human ARG1 was used as the immunogen for the Arginase antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Spécifications
À propos de cet article
This protein is involved in the pathway that synthesizes L-ornithine and urea from L-arginine.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 2-4 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Optimal dilution of the Arginase antibody should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 min
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: ARG1
Clonality: Monoclonal
Clone: ARG1/1126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human