Anti-PRKAA1/PRKAA2 Rabbit Polyclonal Antibody
PRSI14-595
New Product
- Antibody type:Primary
- Antigen name:protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1/PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:5562;5563
- Antigen synonyms:AMPK|HMG-CoA redustase kinase|VEGFR2|AMPK-alpha1|PRKAA1|AAPK1|AMPKa1|AMPK alpha-1 chain|FLK1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of human PRKAA1/PRKAA2
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: PRKAA1/PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat