Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 660)
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 660
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- AATB100210G0
- AATB100210G1
- Antigen symbol:
- CD2
- CD2
- Antigen name:
- CD2 antigen
- CD2 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 660
- iFluor® 660
- Clone:
- RPA-2.10
- RPA-2.10
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
Frequently Bought Together