Anti-GRIN1 Rabbit Polyclonal Antibody
Catalog # PRSI22-753
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 1
- Antigen symbol:GRIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2902
- Antigen synonyms:NMDAR1|glutamate receptor|glutamate [NMDA] receptor subunit zeta 1|ionotropic|subunit zeta-1|NR1|NMDA receptor 1|N-methyl D-aspartate 1|NMDA1|GRIN1|N-methyl-D-aspartate receptor channel
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 600-700 of human GRIN1 (NP_015566.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat