Anti-LONRF3 Rabbit Polyclonal Antibody
Catalog # PRSI59-385
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:LON peptidase N-terminal domain and RING finger protein 3
- Antigen symbol:LONRF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:677 - 704 aa
- Form:Liquid
- Gene ID:79836
- Modification:None
- Molecular weight:84 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,32 mg/ml
- Shipping temperature:4°C
- Immunogen:This LONRF3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 677 - 704 amino acids from the C-terminal region of human LONRF3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: LONRF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 677-704
Host: Rabbit
Isotype: Ig
Reactivity: Human