Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant SOD2 (from Cells)
Human Recombinant SOD2 (from Cells)
Catalog # PRSI91-789
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant SOD2 (from Cells)
Catalog # PRSI91-789
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    SOD2|Mn-SOD|Superoxide Dismutase [Mn] Mitochondrial
  • Protein/peptide name:
    SOD2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23,24 kDa
  • Sequence:
    Lys25-Lys222
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM Tris, 150 mM NaCl, pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Superoxide Dismutase (SOD2) belongs to the iron/manganese superoxide dismutase family. SOD2 is a mitochondrial matrix protein that forms a homotetramer and binds one manganese ion per subunit. SOD2 transforms toxic superoxide, a byproduct of the mitochondrial electron transport chain into hydrogen peroxide and diatomic oxygen. It is reported that oxidative stress plays an essential role in the development of breast cancer, while SOD2 is one of the primary enzymes that directly convert potential harmful oxidizing species to harmless metabolites.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays.

Caution: For research use only.