Anti-PIK3AP1 Rabbit Polyclonal Antibody
PRSI56-700
New Product
- Antibody type:Primary
- Antigen name:Phosphoinositide 3-kinase adapter protein 1
- Antigen symbol:PIK3AP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acid range = 639-668
- Form:Liquid
- Gene ID:Q6ZUJ8
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:90 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PIK3AP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 639-668 amino acids from the C-terminal region of human PIK3AP1
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
PIK3AP1 is involved in the activation of phosphoinositide 3-kinase (PI3K) in B-cells and in natural killer (NK) cells. Couples B-cell antigen receptor (BCR) to PI3K activation by providing a docking site for the PI3K subunit PIK3R1, which contributes to B-cell development. Seems to have a complementary role with CD19 in PI3K activation (By similarity). May be involved in the survival of mature B cells via activation of REL (By similarity).
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: PIK3AP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human