Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Catalog # PRSI76-614
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:UCHT2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P06127
- Antigen synonyms:CD5
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:25 Tests
Specifications
About this item
The UCHT2 monoclonal antibody specifically reacts with human CD5, a 67 kDa type 1 transmembrane glycoprotein. CD5 is expressed on mature T cells, a subset of B cells, and peripheral blood dendritic cells. B cells that are CD5+ produce mostly IgM polyreactive antibodies. Its ligand is CD72, which is involved in T, B cell proliferation and interaction. The UCT2 antibody also recognizes non-human primate CD5.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human