Anti-Cd40 Hamster Monoclonal Antibody [clone: HM40-3] (FITC (Fluorescein Isothiocyanate))
PRSI76-030
New Product
- Antibody type:Primary
- Antigen name:CD40 antigen
- Antigen symbol:Cd40
- Clonality:Monoclonal
- Clone:HM40-3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgM kappa
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:P27512
- Antigen synonyms:Cd40
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,5 mg
The HM40-3 monoclonal antibody reacts with mouse CD40, a 48 kDa type I glycoprotein known as Bp50. CD40 is part of the TNFR family and expressed on a subset of T cells, B cells, dendritic cells, and macrophages. It has been shown to be involved in the protection of B cells from apoptosis, B cell differentiation, co-stimulation, and isotype class-switching. The HM40-3 antibody blocks the binding of CD40L (CD154) and stimulates B and dendritic cell proliferation.
Type: Primary
Antigen: Cd40
Clonality: Monoclonal
Clone: HM40-3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgM kappa
Reactivity: Mouse;Rat