Order Entry
Belgium
ContactUsLinkComponent
Anti-Ly76 Rat Monoclonal Antibody [clone: TER-119] (FITC (Fluorescein Isothiocyanate))
Anti-Ly76 Rat Monoclonal Antibody [clone: TER-119] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-078
Supplier:  ProSci Inc.
New Product
Anti-Ly76 Rat Monoclonal Antibody [clone: TER-119] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-078
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lymphocyte antigen 76
  • Antigen symbol:
    Ly76
  • Clonality:
    Monoclonal
  • Clone:
    TER-119
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    Ly76
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

The TER-119 monoclonal antibody specifically binds to the mouse 52 kDa Glycophorin A expressed on the erythroid lineage cells, from the early proerythroblast to mature erythrocyte stages, in the fetal liver, adult bone marrow, newborn liver, yolk sac, and adult lymphoid organs. The TER-119 antibody is used, together with other antibodies, as a marker for hematopoietic lineages cells.

Type: Primary
Antigen: Ly76
Clonality: Monoclonal
Clone: TER-119
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse