Anti-Cxcr4 Rat Monoclonal Antibody [clone: 2B11] (PE (Phycoerythrin))
Catalog # PRSI76-872
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) receptor 4
- Antigen symbol:Cxcr4
- Clonality:Monoclonal
- Clone:2B11
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P70658
- Antigen synonyms:Cxcr4
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,05 mg
Specifications
About this item
The 2B11 monoclonal antibody specifically reacts with mouse CD184 (CXCR4or Fusin), a seven-transmembrane G-protein-coupled receptor. It is expressed by astrocytes, B cells, T cells, monocytes, neutrophils, neurons, and hematopoietic cell types. CD184 is a coreceptor for X4 HIV-1.The monoclonal 2B11 antibody recognizes the amino terminus of CD184 and has functional activity.
Type: Primary
Antigen: Cxcr4
Clonality: Monoclonal
Clone: 2B11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse