Anti-THY1 Mouse Monoclonal Antibody [clone: 5E10] (PE (Phycoerythrin))
Catalog # PRSI76-068
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Thy-1 cell surface antigen
- Antigen symbol:THY1
- Clonality:Monoclonal
- Clone:5E10
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P04216
- Antigen synonyms:THY1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The 5E10 monoclonal antibody specifically reacts with human CD90, also known as thymus cell antigen-1 (Thy-1). CD90 is a 25 to 37 kDa GPI-anchored protein is the smallest member of the Ig superfamily. CD90 is expressed on thymocytes, neurons, mesenchymal stem cells, hematopoietic stem cells, NK cells, and follicular dendritic cells and plays a role in inflammation, metastasis, apoptosis, and nerve regeneration. The 5E10 antibody can be used for the enriching high proliferative potential colony-forming cells (HIPP-CFC).
Type: Primary
Antigen: THY1
Clonality: Monoclonal
Clone: 5E10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human