Anti-Emr1 Rat Monoclonal Antibody [clone: BM8.1]
PRSI76-052
New Product
- Antibody type:Primary
- Antigen name:EGF-like module containing, mucin-like, hormone receptor-like sequence 1
- Antigen symbol:Emr1
- Clonality:Monoclonal
- Clone:BM8.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:Q61549
- Antigen synonyms:Emr1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography.
- Pk:0,1 mg
The BM8.1 monoclonal antibody specifically binds to the Mouse125 kDa F4/80 antigen, expressed by most mature macrophages. F4/80 is a transmembrane protein used as a marker of macrophages, although it is also expressed on Kupffer and Langerhans cells. The expression of F4/80 antigen is upregulated on bone marrow cells stimulated in vitro with the macrophage colony stimulating factor. The F4/80 antigen is a requirement for the induction of CD8 T cells-mediated peripheral tolerance.
Type: Primary
Antigen: Emr1
Clonality: Monoclonal
Clone: BM8.1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse