Order Entry
Belgium
ContactUsLinkComponent
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (APC (Allophycocyanin))
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (APC (Allophycocyanin))
  PRSI76-627
New Product
Anti-CD27 Mouse Monoclonal Antibody [clone: O323] (APC (Allophycocyanin))
  PRSI76-627
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD27 molecule
  • Antigen symbol:
    CD27
  • Clonality:
    Monoclonal
  • Clone:
    O323
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P26842
  • Antigen synonyms:
    CD27
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    25 Tests

 

 

The O323 monoclonal antibody specifically binds to the human CD27, which is a lymphocyte-specific member of the TNF/NGF-R family, expressed on a subset of human thymocytes and the majority of the mature T cells. CD27 is upregulated by T lymphocytes stimulation, and binds to CD70, enhancing the interaction between T and B lymphocytes. CD27 is a memory marker of B cells.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human