Anti-GRIN2D Rabbit Polyclonal Antibody
Catalog # PRSI13-418
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:glutamate receptor, ionotropic, N-methyl D-aspartate 2D
- Antigen symbol:GRIN2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:2906
- Antigen synonyms:GluN2D|glutamate receptor|NMDAR2D|ionotropic|N-methyl-D-aspartate receptor subunit 2D|EB11|GRIN2D|N-methyl D-aspartate 2D|estrogen receptor binding CpG island
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 330-480 of human GRIN2D (NP_000827.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GRIN2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat