Anti-RET Rabbit Polyclonal Antibody
PRSI63-125
- Antibody type:Primary
- Antigen name:Proto-oncogene tyrosine-protein kinase receptor Ret
- Antigen symbol:RET
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:1063-1093 aa
- Form:Liquid
- Gene ID:5979
- Modification:None
- Molecular weight:124 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This RET antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1063-1093 amino acids from the C-terminal region of human RET.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: RET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 1063-1093
Host: Rabbit
Isotype: Ig
Reactivity: Human