Anti-ARHGEF33 Rabbit Polyclonal Antibody
PRSI60-428
New Product
- Antibody type:Primary
- Antigen name:Rho guanine nucleotide exchange factor 33
- Antigen symbol:ARHGEF33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:753 - 780 aa
- Form:Liquid
- Gene ID:100271715
- Modification:None
- Molecular weight:95 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,32 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ARHGEF33 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 753 - 780 amino acids from the C-terminal region of human ARHGEF33.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ARHGEF33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 753-780
Host: Rabbit
Isotype: Ig
Reactivity: Human