Anti-JAM2 Rabbit Polyclonal Antibody
PRSI24-009
New Product
- Antibody type:Primary
- Antigen name:junctional adhesion molecule 2
- Antigen symbol:JAM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:58494
- Antigen synonyms:VEJAM|CD322|JAM2|JAMB|vascular endothelial junction-associated molecule|C21orf43|OTTHUMP00000096100|VE-JAM|junctional adhesion molecule B|junctional adhesion molecule 2|UNQ219/PRO245|PRO245|JAM-B|JAM-IT/VE-JAM
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 29-238 of human JAM2 (NP_067042.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: JAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human