Order Entry
Belgium
ContactUsLinkComponent
Collagen I (from Rat Tail Tendons)
Collagen I (from Rat Tail Tendons)
Catalog # ENZOALX5224400050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Collagen I (from Rat Tail Tendons)
Catalog # ENZOALX5224400050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Source:
    Tail tendon
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <100 EU/mg purified protein (LAL test)
  • Protein synonyms:
    COL1
  • Protein/peptide name:
    Collagen I
  • Purity:
    ≥90% (SDS-PAGE)
  • Molecular weight:
    235, 215, 130, 115 kDa
  • Concentration:
    100 µg/ml
  • Formulation:
    Sterile liquid, in 60% ethanol
  • Shipping temperature:
    20

Specifications

About this item

Collagen is the main component in connective tissue and helps to provide support for tissues. It is made up of several classes, with Type 1 collagen being the most common.

Type 1 collagen has a herterotrimeric triple helical structure made up of two alpha-1(I) and one alpha-2(I) chains that form into elongated fibrils which are extremely strong. These fibrils can be found in skin, tendons, ligaments, and other connective tissues.

Type 1 collagen has been shown to be useful as a substrate that promotes cell growth and proliferation. Under acidic conditions the protein is soluble, however by raising the temperature and pH the solution forms into a solid gel that can be useful for cellular studies. It can also be dried to form a thin layer on solid surfaces such as plates, slides, or coverslips to aid in cell attachment.