Order Entry
Belgium
ContactUsLinkComponent
Trypsin
Trypsin
Supplier:  Roth Carl
Certificates
Trypsin
Supplier:  Roth Carl

Specifications

  • CAS No.:
    9002-07-7
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00082094
  • Enzyme name:
    Trypsin
  • Purity:
    2X crystallized

Specifications

Frequently Bought Together