Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Trypsin
Trypsin
A3964.0005
:
PanReac AppliChem
:
Trypsin
A3964.0005
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
5 g
CAS No.:
9002-07-7
Storage conditions:
Refrigerator
MDL No.:
MFCD00082094
Enzyme name:
Trypsin
Enzyme activity:
2.500 U/mg