Order Entry
Belgium
ContactUsLinkComponent
 
Starch 1% in aqueous solution
  FFCHFF2431000
 :  
Starch 1% in aqueous solution
  FFCHFF2431000
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

 :
 :162.14 g/mol
 :Ambient
 :232-679-6
 :9005-25-8
 :(C₆H₁₀O₅)n
 :MFCD00082026