Starch 0.5% in aqueous solution stabilised
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:9005-25-8
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-679-6
Frequently Bought Together