Mercury(II) nitrate 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP, Supelco®
Catalog # 1.09143.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.02 g/cm3 (20 °C)
- pH:1 (H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
Synonyms: Mercury dinitrate, Mercuric nitrate
:
:324.60 g/mol
Density:1.025 g/cm³ (25 °C)
UN:3289
ADR:6.1 (8),II
:M-Clarity™ quality level = MQ100
EINECS:233-152-3
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:10045-94-0
Formula:HgN₂O₆
MDL Number:MFCD00011038