Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenol red

Phenol red

50 Tests
Catalog # PALIPM130
Supplier:   Palintest
Phenol red
Catalog # PALIPM130
Supplier:  Palintest
CAS Number:  143-74-8
Phenol red

50 Tests

CAS Number:  143-74-8
Price
20,60 €
Log in for availability
1 * 50 Tests
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Merck Index:13,07329
  • Melting Pt:> 300 °C
  • Storage Temperature:Ambient
  • EINECS:205-609-7

Documents

Specifications

Specifications

  • Pk:
    50 Tests

Specifications