Order Entry
Belgium
ContactUsLinkComponent
Phenolphthalein 1% in ethanol Reag. Ph. Eur. indicator, Fluka™
Phenolphthalein 1% in ethanol Reag. Ph. Eur. indicator, Fluka™
Catalog # 34607-30ML
CAS Number:  
undefined
Phenolphthalein 1% in ethanol Reag. Ph. Eur. indicator, Fluka™
Catalog # 34607-30ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    30 mL
  • Seq:
    2345

Specifications

Specification Test Results

  • Test of sensitivity:
    complying
  • Change range (pH 7.2-10.0):
    complying

Specification Test Results

About this item

 :
 :318.33 g/mol
Boiling Pt:? 78 °C (1013 hPa)
Density:~ 0,85 g/cm³ (20 °C)
Flash Pt:14 °C
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913