Order Entry
Belgium
ContactUsLinkComponent
Phenolphthalein 0.375% in methanol
Phenolphthalein 0.375% in methanol
Supplier:  Merck
Certificates
undefined
Phenolphthalein 0.375% in methanol
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Flash Point:
    11 °C
  • Explosion Limit:
    5.5 - 36.5 %(V)
  • Density:
    0.80 g/cm3 (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together