Order Entry
Belgium
ContactUsLinkComponent
Potassium dihydrogen phosphate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1069601
Potassium dihydrogen phosphate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1069601
Catalog # 87901.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium dihydrogen phosphate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1069601
Catalog # 87901.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :136.09 g/mol
Storage Temperature:Ambient
EINECS:231-913-4
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401