Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate 0,5% in aqueous solution Reag. Ph. Eur. 1069502
Toxic
Health
Potassium dichromate 0,5% in aqueous solution Reag. Ph. Eur. 1069502
Catalog # 87900.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Potassium dichromate 0,5% in aqueous solution Reag. Ph. Eur. 1069502
Catalog # 87900.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :294.18 g/mol
Storage Temperature:Ambient
EINECS:231-906-6
UN:3287
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367