Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate 1/24 mol/l (0.25 N) in aqueous solution, Supelco®
Health
Potassium dichromate 1/24 mol/l (0.25 N) in aqueous solution, Supelco®
Catalog # 1.09118.1000
Supplier:  Merck
CAS Number:  
Health
Potassium dichromate 1/24 mol/l (0.25 N) in aqueous solution, Supelco®
Catalog # 1.09118.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.01 g/cm3 (20 °C)
  • pH:
    3.8 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :294.18 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
EINECS:231-906-6
UN:3287
 :GHS08
Storage Temperature:Ambient
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367