Rhodamine 6G ~95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride
CAS Number:989-38-8
:
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00012665
:479.02 g/mol
Boiling Pt:? 200 °C (1013 hPa)
Melting Pt:215 °C
Density:1,3 g/cm³
:M-Clarity™ quality level = MQ200
EINECS:213-584-9
:GHS05
:GHS06
:GHS09
Storage Temperature:Room Temperature
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility (colour):1 mg/ml in water
- Solubility (method):1 mg/ml in water
- Carbon content:66.5 - 74.0 %
- Nitrogen content:5.0 - 6.2 %
- Dye content:~ 95 %
- Wavelength (1) (UV):~ 525 nm
- Extinction (1%/1 cm) (1) (UV):≥ 1700
- Solvent (UV):Water
Specification Test Results
Frequently Bought Together