ContactUsLinkComponent
Methyl orange 0.1% in aqueous solution Reag. Ph. Eur. indicator, Fluka™
Methyl orange 0.1% in aqueous solution Reag. Ph. Eur. indicator, Fluka™
  34576-500ML
 :  
Methyl orange 0.1% in aqueous solution Reag. Ph. Eur. indicator, Fluka™
  34576-500ML
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Test of sensitivity:
    complying
  • Change range (pH 3,0-4,4):
    complying
 :
 :327.34 g/mol
 :0.970 g/cm3 (20 °C)
 :39 °C
 :Ambient
 :208-925-3
 :1170
 :3,III
 :GHS02
 :547-58-0
 :C₁₄H₁₄N₃NaO₃S
 :MFCD00007502