Order Entry
Belgium
ContactUsLinkComponent
 
Bromophenol blue 0.04% (w/w) in aqueous solution, Sigma-Aldrich®
  318744-500ML
 :  
undefined
Bromophenol blue 0.04% (w/w) in aqueous solution, Sigma-Aldrich®
  318744-500ML
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance (Colour):
    Purple
  • Appearance (Form) :
    Liquid
  • Wave Length
    In Water:
    307 - 313 nm
  • Extinction Coefficient:
    ≥14500
  • Wave Length:
    377 - 383 nm
  • Visual Interval
    pH 3.0 - Yellow with Slight Green Tint:
    ≥75000
  • Visual Interval
    pH 3.4 - Green:
    Pass
  • Visual Interval
    pH 4.6 - Blue:
    Pass
  • undefined:
    Pass

 

 

 :
 :669.97 g/mol
 :0.985 g/ml (25 °C)
 :M-Clarity™ quality level = MQ200
 :Ambient
 :204-086-2
 :115-39-9
 :C₁₉H₁₀Br₄O₅S
 :MFCD00005875