Order Entry
Belgium
ContactUsLinkComponent
Bromocresol green sodium salt 0.5 g/l in ethanol 20% (v/v) Reag. Ph. Eur.
Bromocresol green sodium salt 0.5 g/l in ethanol 20% (v/v) Reag. Ph. Eur.
Catalog # FFCHFF0180050
Supplier:  FF CHEMICALS
CAS Number:  
Bromocresol green sodium salt 0.5 g/l in ethanol 20% (v/v) Reag. Ph. Eur.
Catalog # FFCHFF0180050
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
ADR:3,III
 :720 g/mol
Storage Temperature:Ambient
EINECS:263-657-4
UN:1170
CAS Number:62625-32-5
 :67763-24-0
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898