Order Entry
Belgium
ContactUsLinkComponent
Bromocresol green sodium salt 0.04% in aqueous solution, Sigma-Aldrich®
Bromocresol green sodium salt 0.04% in aqueous solution, Sigma-Aldrich®
  318701-500ML
 :  
undefined
Bromocresol green sodium salt 0.04% in aqueous solution, Sigma-Aldrich®
  318701-500ML
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance (Colour):
    Conforms to Requirements
    Dark Green to Dark Blue
  • Appearance (Form):
    Liquid
  • Wavelength:
    613 - 619 nm
  • Extinction Coefficient :
    ≥25000
  • Visual Interval
    pH 3.8 - Yellow:
    Conforms to Requirements
  • Visual Interval
    pH 4.5 - Green:
    Conforms to Requirements
  • Visual Interval
    pH 5.4 - Blue:
    Conforms to Requirements

 

 

 :
 :0.981 g/ml (25 °C)
 :M-Clarity™ quality level = MQ100
 :720 g/mol
 :Ambient
 :263-657-4
 :62625-32-5
 :67763-24-0
 :C₂₁H₁₃Br₄NaO₅S
 :MFCD00148898