Order Entry
Belgium
ContactUsLinkComponent
Methyl red sodium salt
Methyl red sodium salt
Supplier:  Merck
Certificates
Methyl red sodium salt
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    800 g/l (25 °C)
  • Molar Mass:
    291.29 g/mol
  • Bulk Density:
    360 kg/m3
  • pH:
    8.2 (10 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Frequently Bought Together