Methyl red, pure indicator
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- CAS Number:493-52-7
- Formula:C₁₅H₁₅N₃O₂
- MDL Number:MFCD00002425
- Molecular Weight:269.3 g/mol
- Merck Index:12,06199
- Melting Pt:178…182 °C
- Density:1,31 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-776-1
3 Options Available Below
Product Details & Documents
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- CAS Number:493-52-7
- Formula:C₁₅H₁₅N₃O₂
- MDL Number:MFCD00002425
- Molecular Weight:269.3 g/mol
- Merck Index:12,06199
- Melting Pt:178…182 °C
- Density:1,31 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-776-1
Specifications for Product Family
Specification Test Results
- Appearance:Dark red to brown to purple to violet Powder or crystals
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Solubility:(0.1 % in ethanol) Clear
- Specific extinction E (1%/1cm):≥650 (430 nm, in buffer)
- Visual transition interval:from pH 4.2 (pink or red) to pH 6.2 (yellow)



