Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) substrate

3,3',5,5'-Tetramethylbenzidine (TMB) substrate

50 mL
Catalog # ENZOADI801805
Supplier:   Enzo Life Sciences
3,3',5,5'-Tetramethylbenzidine (TMB) substrate
Catalog # ENZOADI801805
Supplier:  Enzo Life Sciences
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) substrate

50 mL

CAS Number:  54827-17-7
Price
110,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

HRP substrate.

  • Used in ELISA

This TMB Substrate is intended for use as a colourimetric HRP-substrate for immunoassay development. Each bottle is sufficient for use with 5×96 wells microtiter plates.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pk:
    50 mL

Specifications